2-(Trifluoromethyl)cyclohexanone ≥97%

Supplier: Matrix Scientific

005497-1G 005497-5G
101789-058EA 169.23 USD
101789-058 101789-060
2-(Trifluoromethyl)cyclohexanone ≥97%
2-(Trifluoromethyl)cyclohexanone

Hazard: Flammable

Formula: C₇H₉F₃O
MW: 166.14 g/mol
Boiling Pt: 71…73 °C (12 mmHg)
MDL Number: MFCD00102125
CAS Number: 56734-74-8
UN: 1224
ADR: 3 III

Order Now

Specification Test Results

Boiling point 74°C/18mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR