4,6-Dichlorosalicylaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloro-6-hydroxybenzaldehyde

005123-1G
101788-290EA 90.77 USD
101788-290
4,6-Dichlorosalicylaldehyde ≥97%
4,6-Dichlorosalicylaldehyde

Hazard code(s): Irritant, Air Sensitive

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Melting Pt: 51…53 °C
MDL Number: MFCD00016590
CAS Number: 78443-72-8

Order Now

Specification Test Results

Melting point 51-53°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR