2,2,2-Trifluoroethyl methanesulfonate ≥97%

Supplier: Matrix Scientific

003705-25G 003705-100G
101786-100EA 112.31 USD
101786-100 101786-102
2,2,2-Trifluoroethyl methanesulfonate ≥97%
2,2,2-Trifluoroethyl methanesulfonate

Hazard: Irritant

Formula: C₃H₅F₃O₃S
MW: 178.130 g/mol
Density: 1.52
MDL Number: MFCD00025058
CAS Number: 25236-64-0

Order Now

Specification Test Results

Density 1.52
ND20 1.367


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR