2,3,5,6-Tetrafluorothiophenol ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Mercapto-2,3,5,6-tetrafluorobenzene, 2,3,5,6-Tetrafluorobenzenethiol, 2,3,5,6-Tetrafluorobenzene-1-thiol, 2,3,5,6-tetrafluorothiophenol

003486-5G
101785-774EA 209.23 USD
101785-774
2,3,5,6-Tetrafluorothiophenol ≥97%
2,3,5,6-Tetrafluorothiophenol

Hazard code(s): Irritant, Stench

Formula: C₆H₂F₄S
MW: 182.14 g/mol
Boiling Pt: 152 °C (1013 hPa)
MDL Number: MFCD00004829
CAS Number: 769-40-4
EINECS: 212-210-1
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Boiling point 152-153°C
ND20 1.4865

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR