Allyl-1H,1H-perfluorooctyl ether ≥97%

Supplier: Matrix Scientific

001928-5G
101782-866EA 446.15 USD
101782-866
Allyl-1H,1H-perfluorooctyl ether ≥97%
Allyl-1H,1H-perfluorooctyl ether

Hazard: Irritant

Formula: C₁₁H₇F₁₅O
MW: 440.15 g/mol
Boiling Pt: 178°C
MDL Number: MFCD00042149
CAS Number: 812-72-6

Order Now

Specification Test Results

Boiling point 178°C
ND20 1.3222

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR