1-Iodo-4,4,4-trifluorobutane ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Iodo-1,1,1-trifluorobutane, 1,1,1-Trifluoro-4-iodobutane

001897-5G
101782-820EA 150.77 USD
101782-820
1-Iodo-4,4,4-trifluorobutane ≥97%
1-Iodo-4,4,4-trifluorobutane

Hazard code(s): Irritant, Light Sensitive

Formula: C₄H₆F₃I
MW: 237.99 g/mol
Boiling Pt: 126…127 °C
Density: 1.85 g/cm³
MDL Number: MFCD00041543
CAS Number: 461-17-6

Order Now

Specification Test Results

Boiling point 126-127°C
ND20 1.4326

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR