indane-5-sulfonyl chloride

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydro-1H-indene-5-sulphonyl chloride

011907-1G
101780-098EA 215.38 USD
101780-098
indane-5-sulfonyl chloride
indane-5-sulfonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₉H₉ClO₂S
MW: 216.679 g/mol
Boiling Pt: 180 °C (16 Torr)
Melting Pt: 46 °C
MDL Number: MFCD05237217
CAS Number: 52205-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR