Undecafluoro(2-methyl-3-oxahexanoyl)fluoride ≥97%

Supplier: Matrix Scientific

Synonyms: Perfluoro(2-methyl-3-oxahexanoyl) fluoride

004968-5G
101777-864EA 126.15 USD
101777-864
Undecafluoro(2-methyl-3-oxahexanoyl)fluoride ≥97%
Undecafluoro(2-methyl-3-oxahexanoyl)fluoride

Hazard: Corrosive

Formula: C₆F₁₂O₂
MW: 332.05 g/mol
Boiling Pt: 54-56°C
MDL Number: MFCD00054657
CAS Number: 2062-98-8

Order Now

Specification Test Results

Boiling point 54-56°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR