2-Fluoro-3-(trifluoromethyl)benzylamine ≥97%

Supplier: Matrix Scientific

002299-5G
101775-960EA 227.69 USD
101775-960
2-Fluoro-3-(trifluoromethyl)benzylamine ≥97%
2-Fluoro-3-(trifluoromethyl)benzylamine

Hazard: Corrosive/Store in Freezer

Formula: C₈H₇F₄N
MW: 193.14 g/mol
Boiling Pt: 193 °C
Density: 1.321
Flash Pt: 79.6 °C
MDL Number: MFCD00061161
CAS Number: 239135-49-0

Order Now

Specification Test Results

Boiling point 193 deg. C
Density 1.321

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR