Phusion GC Buffer Pack, New England Biolabs
Supplier: New England Biolabs (NEB)
B0519S
101640-766EA 29 USD
101640-766
Phusion GC Buffer Pack, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
New England Biolabs supplies a unique 5X reaction buffer and supplemental reagents for use with Thermo Scientific® Phusion High-Fidelity DNA Polymerase and associated products, all of which are manufactured and quality controlled at New England Biolabs.