Green Supreme HEPA Vac, Atrix
Supplier: Atrix International
VACGRNS
101640-734EA 434.58 USD
101640-734
Green Supreme HEPA Vac, Atrix
Vacuum Cleaners
The Green Supreme 1 Gallon HEPA dry particulate vacuum is ideal for the RRP Contractor
Replacement Filters are designed to capture fumes, atmospheric dust, black toner and all color toner and other ultra-fine particulate. Compatible with 3M vacuums.
With its self contained HEPA filter, hose plugs and filter plugs, the VACGRNS makes filter changes, transporting and storing safe. Vacuum is compact and portable. HEPA Efficiency: 99.97% @ .3 microns.
Información para pedidos: Includes self-storing utensils (cover insert), 6' ESD Safe hose, two hose plugs, 16" ESD gooseneck, two 18"More Product Information