Alere EZ Split Key® Cup and Key Cup A.D., CLIA Waived Drug Test Kits, Alere™ Toxicology
Supplier: Alere
DUD-157-012-019 DUD-157-023-019 DOA-3257-019 DOA-1167-019 DUA-147-012-019 DOA-1237-019 DOA-1247-019 DOA-1257-019 DOA-2257-019
10154-068BX 188.54 USD
10154-068 10154-070 10154-078 10154-080 10154-082 10154-084 10000-708 10000-710 10000-712
Alere EZ Split Key® Cup and Key Cup A.D., CLIA Waived Drug Test Kits, Alere™ Toxicology
Drug Tests
Rapid Drug Screen for the simultaneous, qualitative detection of single or multiple drug metabolites in human urine
Stand up cup that detects drug metabolites in urine with results in 5 minutes. Split specimen cup with key activation. Cup and temperature strip included, SVT option available.
Room temperature storage, long shelf-life.
Rapid screen for a collection of illicit and prescription drugs. Each kit contains materials for 25 drug tests. Rapid Drug Screens can include Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP), Cocaine (COC),More Product Information