Alere iCup® and iCup® A.D., Moderately Complex/For Professional Use Drug Test Kits, Alere Toxicology
Supplier: Alere
I-DUA-157-034 I-DUA-147-012 I-DUA-157-023 I-DUA-167-012 I-DUA-157-013 I-DOA-3137 I-DOA-177-031 I-DUD-187-013 I-DUE-167-342 I-DUE-187-071 I-DOA-1107-051 I-DOA-1137-011 I-DUA-167-022 I-DUA-167-291 I-DUD-197-014 I-DUD-1107-012 I-DUE-1107-141 I-DUE-1107-291 I-DOA-197-041 I-DUE-1127-022
89171-320PK 307.5 USD
89171-320 89423-370 89423-372 89423-374 10154-054 10154-056 10154-058 10154-060 10154-062 10154-064 10154-066 10000-520 10000-522 10000-524 10000-526 10000-528 10000-530 10000-532 10000-534 10000-536 10000-700 10000-702
Alere iCup® and iCup® A.D., Moderately Complex/For Professional Use Drug Test Kits, Alere Toxicology
Drug Tests
Rapid Drug Screen for the simultaneous, qualitative detection of single or multiple drug metabolites in human urine
Stand up cup that detects drug metabolites in urine with results in 5 minutes. Cup and temperature strip included, SVT option available.
Room temperature storage, long shelf-life.
Rapid screen for a collection of illicit and prescription drugs. Each kit contains materials for 25 drug tests. Rapid Drug Screens can include Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP), Cocaine (COC), Methamphetamine (mAMP), Opiates (OPI) and (MOP),More Product Information