VWR® Mini Hotplate 120 V
Supplier: VWR International
10153-300 10153-312
10153-300EA 308.25 USD
10153-300 10153-312
VWR® Mini Hotplate 120 V
Heaters Hot Plates
Rugged, compact hotplates with an ergonomic design are ideal for educational labs and can heat up to 1000 mL of water.
  • Rugged 5x5" aluminum top plate
  • Boils 300 mL of water in 16 minutes
  • Built-in support rod holder
  • Five-Year Warranty on Parts and Labor
Durable 5x5" cast aluminum top plate will not chip or crack, and provides a uniform heating surface
The fixed temperature hotplate features an illuminated rocker switch to activate the preset fixed temperature of 375°C.
The built-in support rod holder with locking knob accepts an optional supportMore Product Information