Laboratory Freezer Rooms, Precision Controlled Environments, Darwin Chambers Company
Supplier: Darwin Chambers Company
VWLABR1NN08X08 VWLABR1NN10X12 VWLABR1NN10X20
10153-252EA 53007.35 USD
10153-252 10153-254 10153-256
Laboratory Freezer Rooms, Precision Controlled Environments, Darwin Chambers Company
Prefabricated Rooms
Darwin Laboratory Freezer Rooms are designed for maximum reliability, efficiency, and stability.
  • Modular rooms, featuring "tongue-in-groove" construction
  • All major components are pre-wired, pre-brazed, and pre-assembled, ready for quick onsite assembly, defrost requirement minimized by system design
  • Rooms are constructed using 10.2 cm (4") insulated panels, R Value 37+, with white embossed aluminum interior surface finishes and white embossed galvanized steel exterior finishes
  • Include a 10.2 cm (4") insulated smooth aluminum or stainless steel floor,
More Product Information