Canine Serum, ImmunoReagents
Supplier: Immunoreagents
SP-128-VX10 SP-128-VX2
10152-194EA 209.09 USD
10152-194 10152-196
Canine Serum, ImmunoReagents
Sera (Serum)
ImmunoReagents’ normal canine serum is used as a blocking agent or as a control for immunoassays
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulin s from various species cross absorbed to provide high specificity. 
All serum is obtained from healthy, disease free animals. Serum isMore Product Information