Anti-IgG Horse Polyclonal Antibody

Supplier: Immunoreagents
EQ-003-C
10151-052EA 142.75 USD
10151-052
Anti-IgG Horse Polyclonal Antibody
Antibodies
  • Origin: US

Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Horse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR