Anti-IgG Human Polyclonal Antibody

Supplier: Immunoreagents
HU-003-C
10151-032EA 191.86 USD
10151-032
Anti-IgG Human Polyclonal Antibody
Antibodies
  • Origin: US

Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR