HotStart Taq DNA Polymerase, IBI Scientific
Supplier: Avantor
10146-876EA 225.03 USD
10146-876 10146-878 10146-902 10146-904
HotStart Taq DNA Polymerase, IBI Scientific
Nucleic Acid Reagents End-point PCR Enzymes and Kits
IBI Taq HotStart DNA Polymerase is a chemically modified hot start enzyme excellent for preventing or minimizing non-specific DNA amplification
Under standard hot start conditions (94⁰C in TRIS w/pH 8-9) IBI Taq HotStart regains both the 5’→3’ polymerase and 5’→3’ exonuclease activity within two minutes. IBI Taq HotStart suppresses enzyme activity at room temperature with near full recovery of enzyme activity following activation. Our proprietary purification process also produces a contaminant- and nuclease-free enzyme that offers high-copy PCR products from low concentration template or enzyme.
The 10X PCR Buffer contains 100mM Tris-HCl (pH 8.4), 500mM KCl, with or without 15mM MgCl2.
The Storage Buffer contains 20mM HEPES (pH 7.9), 100mM KCl, 0.1mM EDTA, 0.5mM PMSF, 1mM DTT, 50% (v/v) glycerol, stabilizers.
The novel modifying reagent creates a faster activation time, an improvement over similar chemically modified hot start enzymes. The alkaline activation condition makes for greater activity and better specific amplification which minimizes primer-dimers. The superior stability of IBI Taq HotStart also means a longer shelf life.
Certificaciones: Contaminant- and Nuclease-Free.
Información para pedidos: IBI Taq HotStart DNA Polymerase is available in three configurations. The first option is IBI Taq HotStart DNA Polymerase with separate vial(s) of reaction buffer with MgCl2 added. The second option is IBI Taq HotStart DNA Polymerase without any additional buffers. The third option is IBI Taq HotStart DNA Polymerase with separate vial(s) of reaction buffer and a separate vial of MgCl2.
Información de suministro: Shipping conditions Dry Ice