Micro-Touch® Affinity® 3770 Polychloroprene Examination Gloves, Ansell
Supplier: Ansell Healthcare
3770 3771 3772 3773 3774
10146-672BX 53.4 USD
10146-672 10146-674 10146-676 10146-678 10146-680
Micro-Touch® Affinity® 3770 Polychloroprene Examination Gloves, Ansell
Gloves
Micro-Touch Affinity examination gloves provide the fit, comfort, tactile sensitivity and stretchability of natural rubber latex without the risks associated with NRL or glove powder complications.
  • A polymer coating provides excellent donnability for this polychloroprene glove
  • Thickness: 6.3 mil (finger)
Certificaciones: FDA approved for handling chemotherapy drugs.
Información para pedidos: 100 gloves/box; 10 boxes/case.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Disposable/Reusable Disposable
Latex/Latex-free Latex-Free
Powdered/Powder-free Powder-Free
Finish Textured Fingertips
Cuff Style Beaded
Cuff Thickness 4.3 mils
Finger Thickness 6.3 mils
Palm Thickness 5.1 mils
FDA Regulated +