Nickel(II) trifluoromethanesulfonate ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Trifluoromethanesulphonic acid Nickel(II) Salt, Nickel(II) Triflate, Nickel(II) trifluoromethanesulphonate, Nickel triflate

28-1700-1G 28-1700-5G
101442-424EA 58.03 USD
101442-424 101442-426
Nickel(II) trifluoromethanesulfonate ≥98%
Nickel(II) trifluoromethanesulfonate

Metal Triflate

Formula: C₂F₆NiO₆S₂
MW: 356.82 g/mol
MDL Number: MFCD00673740
CAS Number: 60871-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR