Cleanroom NovaWipes, Micronova
Supplier: Micronova
WP4-99IR WP20-99 WPAA-1212 WP2-954 WPAW-1212 WPAA-99 WP2-1012 WP4-99 WP2-1418 WP2-1212 WP2-99 WP6-99H
68400-850CS 2254.76 USD
68400-850 68400-748 94023-890 94023-892 94023-897 10054-530 10142-778 10218-752 94023-895 68400-124 68400-132 76411-422 76411-424 76411-426 68400-141
Cleanroom NovaWipes, Micronova
Wipes Reusable Cleaning Cloths Cloths for Delicate Tasks
A range of absorbent and low-linting wipes to meet every cleaning, sanitizing, polishing and drying application in the cleanroom, support areas, and research suites.
  • Made with cleanroom approved materials
  • Durable, autoclavable, reusable and absorbent
  • Choose from a range of fabrics for specialized cleaning
  • Ideal for parts and surface cleaning, removing tough soils, liquid removal
  • Following laundering, wipes can be used in less critical areas
  • Two-ply wipes feature an inverted hem
  • Hold up to strong disinfectants
NovaCel™ Wipes: Ultra-clean PolyvinylMore Product Information