Tetrabutyltin ≥94%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tetrabutylstannane, Tetra-n-butyltin

93-5049-250G 93-5049-50G
101419-162EA 152.86 USD
101419-162 101419-164
Tetrabutyltin ≥94%
Tetrabutyltin
Formula: C₁₆H₃₆Sn
MW: 347.17 g/mol
Boiling Pt: 145
Melting Pt: -97
Density: 1.0572
Flash Pt: 225 °F
Storage Temperature: Ambient
MDL Number: MFCD00009422
CAS Number: 1461-25-2
EINECS: 215-960-8
UN: 1760
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR