NEB®10-beta Electrocompetent E.coli, New England Biolabs
Supplier: New England Biolabs (NEB)
C3020K
101417-922EA 256 USD
101417-922
NEB®10-beta Electrocompetent E.coli, New England Biolabs
Competent Cells
NEB 10-beta electrocompetent E. coli cells are optimized for high efficiency transformation by electroporation. These cells are ideal for DNA library constructions and all cloning purposes.
  • High efficiency strain ideal for cloning large plasmids and BACs
  • Also available in chemically-competent format (single-use and 200 µl vials)
  • No dry ice surcharge on competent cell shipments
  • Outgrowth medium included
  • Free of animal products