Gram's iodine solution

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Lugol's iodine, Iodine and potassium iodide, Potassium triiodide

26105-04
101411-034EA 46.26 USD
101411-034
Gram's iodine solution
Gram's iodine solution

Brown and Brenn Method reagent for Gram Positive and Gram Negative Bacteria


Gram Negative Bacteria for bacteria stain: Gram + / Gram – bacteria and filaments of nocardia, etc


Stain results - Gram + Bacteria, Nocardia and Actinomyces Filaments: Blue, Gram – Bacteria, Nuclei: Red, Additional tissue elements: Yellow

Formula: I₃K
MW: 419.812 g/mol
MDL Number: MFCD00134128
CAS Number: 12298-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR