Tartrazine 1.5% in aqueous solution

Supplier: Electron Microscopy Sciences

Synonyms: FD and C Yellow No. 5, Filter Yellow, Acid Yellow 23, Sodium 5-oxo-1-(4-sulphonatophenyl)-4-((4-sulphonatophenyl)diazenyl)-4,5-dihydro-1H-pyrazole-3-carboxylate

21741
101410-824EA 53.39 USD
101410-824
Tartrazine 1.5% in aqueous solution
Tartrazine
Formula: C₁₆H₁₂N₄O₉S₂
MW: 534.36 g/mol
MDL Number: MFCD00148908
CAS Number: 1934-21-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cbc869f2767e2943f7d4223ade7f32dc15a63b8&vid=e1691cadc15e8c3b6248f5b8925a8e211d42e713&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR