Sodium metabisulphite 5% aqueous solution

Supplier: Electron Microscopy Sciences

Synonyms: Disodium disulphite, di-Sodium disulphite, Sodium pyrosulphite, Disodiumdisulphite, Sodium disulphite

21175-5
101410-810EA 66.74 USD
101410-810
Sodium metabisulphite 5% aqueous solution
Sodium metabisulphite

Sodium Meta-Bisulfate solution is available in concentrations of 1.0% aqueous, 3.0% aqueous and 5.0% aqueous.

Formula: Na₂S₂O₅
MW: 190.11 g/mol
Melting Pt: 150
MDL Number: MFCD00167602
CAS Number: 7681-57-4
Merck Index: 12,08784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59f60ec761f7ebe73baa05cbe69b20e06dbed6e6&vid=bc750e6aaeb052d58fdcef8a0732a89e62e8015c&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR