Excelsior™ AS Tissue Processors, Epredia™
Supplier: Epredia
A82300001A A82300001HS A82300001ER
10141-592EA 123974.69 USD
10141-592 10141-594 10141-596 10141-598
Excelsior™ AS Tissue Processors, Epredia™
Tissue Processors
Get the superb tissue quality and proven reliability you expect, plus a safer, more efficient working environment, with the Epredia™ Excelsior™ AS Tissue Processor which provides exceptional and enhanced tissue quality with minimal user interaction.
  • Cassette baskets are gently rotated inside the circular chamber
  • More effective agitation than an impeller
  • Reagents can be pre–heated to up to 35°C
  • Minimizes tissue shock and facilitating reagent penetration
  • Alcohol quality measurement enables you to extend your reagent life
  • Provides significant cost savings
More Product Information