M-Vac and M-Vac Jr. Waste Aspiration Systems, Argos Technologies
Supplier: Antyila Scientific
13050-38 13050-39 04396-09 04396-11 04396-12 04396-13 04396-14
89129-566EA 1037.71 USD
89129-566 89129-568 89129-572 10141-550 10141-552 10141-554 10141-556 10141-558
M-Vac and M-Vac Jr. Waste Aspiration Systems, Argos Technologies
Pumps Vacuum Aspiration Systems
Argos M-Vac and M-Vac Jr
UV-resistant systems can be used in laminar hoods. Entire unit is autoclavable at 120°C for 20 minutes ensuring maximum safety. Withstands 700mm Hg. Systems consist of a stainless steel safety stand, 4L or 2L polypropylene vacuum bottle, and a polypropylene cap available with either quick release coupling tubing connectors or traditional barbed fittings (the M-Vac Jr. is available only with the barbed fitting cap). Both units include flexible silicone vacuum tubing that can be used with Pasteur pipettes, serological pipettors or the HandEvac handheld aspirator (More Product Information