RBC Lysis Buffer (10X), Tonbo Biosciences
Supplier: Tonbo Biosciences
TNB-4300-L100
10141-140EA 46.2 USD
10141-140
RBC Lysis Buffer (10X), Tonbo Biosciences
Buffers Extraction Buffers
RBC Lysis Buffer (10X) is a concentrated ammonium chloride-based lysing reagent
The diluted 1X working solution will lyse red blood cells in single cell suspensions with minimal effects on leukocytes. RBC Lysis Buffer (10X) does not contain a fixative so the cells remain viable after red blood cell lysis.