Spray Chamber Helix™ Locking Screw, PerkinElmer
Supplier: PerkinElmer
N0777438
10139-402EA 268.57 USD
10139-402
Spray Chamber Helix™ Locking Screw, PerkinElmer
Fasteners Screws
The spray chamber Helix™ locking screw is used with ICP-MS and ICP-OES series instruments.
Información de suministro: Includes one locking screw.