Titanium High Pressure Capsules, PerkinElmer
Supplier: PERKINELMER U.S. LLC
B0182903
10139-014EA 1566.89 USD
10139-014
Titanium High Pressure Capsules, PerkinElmer
Trays/Dishes/Pots Laboratory Trays/Dishes
High pressure, reusable titanium capsules are designed for use with DSC 4000, DSC 6000, DSC 8000, DSC 8500, Diamond DSC, Pyris 1 DSC, DSC 7, DSC 6, Pyris 6 DSC, and Jade DSC. They use replacement seals (B0185054).
  • Operating range of –170 to 400 °C
  • Capsule volume is 30 µl
  • 150 bar maximum pressure
High pressure capsules are recommended whenever higher pressures are expected. The covers of these capsules contain a bursting disk that allow the sample to escape in the event that pressure exceeds 150 atm. This allows the endothermic signal produced byMore Product Information