Cryofill Dewar Tank for DMA 8000 Dynamic Mechanical Analyzer, PerkinElmer
Supplier: PerkinElmer
N5373009
10138-640EA 10849.63 USD
10138-640
Cryofill Dewar Tank for DMA 8000 Dynamic Mechanical Analyzer, PerkinElmer
Dewars and Cryogenic Vessels
Cooling accessory for the DMA 8000 dynamic mechanical analyzer.
Two options for sub-ambient operation include a 1 l dewar for those needing occasional cooling, and a 50 l dewar for those requiring longer or more frequent sub-ambient experiments.
Información para pedidos: For the 1 l Dewar, please contact your Avantor representative.