Viton Drain Pump Tubing and Connector Kit, PerkinElmer
Supplier: PerkinElmer
N0777455
10136-458EA 973.35 USD
10136-458
Viton Drain Pump Tubing and Connector Kit, PerkinElmer
Tubing Transfer Tubing
This Drain Pump Tubing and Connector Kit, for oils applications, contains Viton® tubing and connectors for the peristaltic pumps in the ASX-260, ASX-520, ASX-520HS, EXR-8, XLR-8, ASX-1400, ASX-1600, and APS-1650.
Enough tubing and connectors are included for two replacements.