Graphite Contact Cylinders, PerkinElmer
Supplier: PerkinElmer
B0504036 B0504035 B3002102 B3002103
97013-420PK 3011.56 USD
97013-420 97013-422 10136-178 10136-180
Graphite Contact Cylinders, PerkinElmer
Tubes Graphite Tubes
The graphite contact cylinders are engineered for a precise fit so that variations in electrical contact, which might affect the analytical performance, are reduced to a minimum.
  • Made from a proprietary blend of base graphite material
  • Produced within tight tolerances to guaranty exact fitment and maximize performance
  • Modified contacts used in applications to minimize condensation and improve analytical performance with difficult matrices (e.g., 2% Ni solutions, edible oils, etc.)
The proper performance, maintenance and serviceability of PerkinElmerMore Product Information