FB Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
K31
10127-472PK 134.61 USD
10127-472
FB Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
(Fastidious Bacteria Broth) For the enriched cultivation of neisseria, haemophilus, streptococcus, corynebacteria and other fastidious bacteria from clinical specimens.
(Fastidious Bacteria Broth) For the enriched cultivation of neisseria, haemophilus, streptococcus, corynebacteria and other fastidious bacteria from clinical specimens.
Información para pedidos: Order by the pack of 20.