VWR® Two-Line Labeling Gun, Black
Supplier: VWR International
10123-976
10123-976CS 202.34 USD
10123-976
VWR® Two-Line Labeling Gun, Black
Printers Label Printers
This two-line labeling gun features an ergonomic handle, reinforced ABS body with steel internal components, easy drop-in label loading, a simple snap-in/pop-out ink roller, and a self-align feed mechanism that provides jam-resistant operation.
  • Autoclavable adhesive withstands extremely high temperatures for up to 20 minutes
  • Easy to replace ink roller
All labels allow the user to dial in two lines of information, with up to eight characters per line to be printed
The accessory easy pop-in/out ink roller is a replacement roller for the VWR® BlackMore Product Information