Human Peripheral Blood Neutrophils, Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
PB011C-1 PB011C-2
101190-080EA 998.08 USD
101190-080 101190-082
Human Peripheral Blood Neutrophils, Charles River Laboratories Cell Solutions
Blood
Human peripheral blood neutrophils are derived from a Dextran red cell sedimented population of cells
Whole blood is collected in HemaCare's FDA-registered collection centers following cGMP and cGTP collection guidelines from healthy human volunteer donors under IRB approved informed consent.
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
Following red cell sedimentation, granulocytes are separated using a density gradient and red cell lysis. Cells areMore Product Information