Mastercycler® nexus X2 PCR Thermal Cycler
Supplier: Eppendorf
6337000027 6336000023 6339000024 6338000020
71003-562EA 10631.34 USD
71003-562 71003-564 71003-566 71003-568 89412-266 89412-268 89412-270 89412-272 89412-274 89412-276 10119-844 10119-846 10119-850 10119-852
Mastercycler® nexus X2 PCR Thermal Cycler
Thermal Cyclers
For a high PCR workload, the Eppendorf® Mastercycler nexus X2 offers efficiency with two blocks (32-well and 64-well), allowing simultaneous runs of two independent protocols. Accommodates a range of consumables up to 0.5 ml tubes. For increased capacity, connect with up to three additional units.
  • Asymmetrically divided block for running different assays in parallel, one 32-well and one 64-well block
  • Intuitive graphic programming
  • Universal block for strips, 0.1, 0.2 and 0.5 ml PCR tubes and fits for divisible PCR plates
  • Small footprint
  • Optional gradient
More Product Information