D-(+)-Maltose monohydrate 92+% (dry basis), Ultrapure

Supplier: Thermo Fisher Scientific

Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate

J18725-A7 J18725-A1
101172-718EA 287.86 USD
101172-718 101175-096
D-(+)-Maltose monohydrate 92+% (dry basis), Ultrapure
D-(+)-Maltose monohydrate
Formula: C₁₂H₂₂O₁₁·1H₂O
MW: 360.32 g/mol
Melting Pt: ∼ 120 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149343
CAS Number: 6363-53-7
EINECS: 200-716-5
Merck Index: 14,5714
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f861d7adf53134a0c485b5942d3be34c4d0b571&vid=16e5c4425a7a8e158a4200470a45483c7ff5781e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Glucose Content (Dry Basis) ≤3.0%
Assay Maltose Content (Dry Basis) ≥92.0%
Heavy Metals (Pb) ≤5 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR