Storage Bins
Supplier: Treston
SBS4-2 SBS4-1015-4ESD SBS4-2ESD SBS4-1015-6 SBS6-3020-4ESD SBS6-3010-6 SBS6-3015-6 SBS6-3020-6 SBS6-3010-4ESD 3015-6 1520-6 1930-6 1015-1 1525-5 1525-6 1930-1 1930-3 1930-5 1940-1 1940-3 1940-5 1940-6 1950-1 1950-3 1950-5 1950-6 3040-3 3040-5 3040-6 3050-3 3050-5 3050-6 3010-1 3010-3 3010-5 3010-6 4010-1 4010-3 4010-5 4010-6 5010-1 5010-3 5010-5 5010-6 3015-1 3015-3 3015-5 4015-1 4015-3 4015-5 4015-6 5015-3 5015-5 5015-6 6015-3 6015-5 6015-6 3020-1 3020-3 3020-5 3020-6 4020-1 4020-3 1015-3 1015-5 1015-6 1520-1 1520-3 1520-5 4020-5 4020-6 5020-1 5020-3 5020-5 5020-6 6020-1 6020-3 6020-5 6020-6 1525-1 1525-3 1015-4ESD 1520-4ESD 1525-4ESD 1930-4ESD 1940-4ESD 1950-4ESD 3040-4ESD 3050-4ESD 30-1L-4ESD
76015-030PK 52.54 USD
76015-030 76015-032 76015-034 76015-060 76015-062 76015-064 76015-066 76015-068 76015-070 10028-005 10028-007 10028-009 10117-695 10117-697 10117-699 10117-701 10117-703 10117-705 10117-707 10117-709 10117-711 10117-713 10117-715 10117-717 10117-719 10117-721 10117-723 10117-725 10117-727 10117-729 10117-731 10117-733 10117-735 10117-737 10117-739 10117-741 10117-743 10117-745 10117-747 10117-749 10117-751 10117-753 10117-755 10117-757 10117-759 10117-761 10117-763 10117-767 10117-769 10117-771 10117-773 10117-775 10117-777 10117-779 10117-781 10117-783 10117-785 10117-787 10117-789 10117-791 10117-793 10117-795 10117-797 10117-819 10117-821 10117-823 10117-825 10117-827 10117-829 10117-863 10117-865 10117-867 10117-869 10117-871 10117-873 10117-875 10117-877 10117-879 10117-881 10117-887 10117-889 10486-477 10496-567 10496-569 10496-571 10682-743 10682-745 10682-747 10682-749 75784-175
Storage Bins
Boxes Storage Boxes
These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.
  • Straight sides ensure an efficient use of space
  • Multiple color options make finding the right item easy
  • Open front provides easy access
  • Corrugated base helps retrieve contents, especially smaller sized and flat objects
Información para pedidos: Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact AvantorMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable