LED Light Fixtures
Supplier: Treston
US-91849010P US-91849006P
10117-574EA 231.25 USD
10117-574 10117-576 10117-578 10117-580 10117-582 10117-584 10117-586 10496-730 10496-732 10496-734
LED Light Fixtures
Lights LED Lights, Lamps and Bulbs
Height and depth adjustable light and balance rails are fitted to the upright tubes and used for suspending lighting units and power tools.
  • Maximum load: 33 lbs.
  • Can hang tools up to 3 lbs.
Información para pedidos: Installation is required. Assembly videos available.
Order Now
SPECIFICATIONS
Environmentally Preferable