Anti-SERPINE1 Rabbit Polyclonal Antibody
Supplier: Prosci
30-286
10109-852EA 760.88 USD
10109-852
Anti-SERPINE1 Rabbit Polyclonal Antibody
Antibodies
SERPINE1 acts as 'bait' for tissue plasminogen activator, urokinase, and protein C. Its rapid interaction with TPA may function as a major control point in the regulation of fibrinolysis.
SERPINE1 antibody can be used for detection of SERPINE1 by ELISA at 1:1562500. SERPINE1 antibody can be used for detection of SERPINE1 by western blot at 1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. FinalMore Product Information