Anti-SLC25A16 Rabbit Polyclonal Antibody
Supplier: Prosci
29-952
10109-262EA 694.55 USD
10109-262
Anti-SLC25A16 Rabbit Polyclonal Antibody
Antibodies
SLC25A16 is a protein that contains three tandemly repeated mitochondrial carrier protein domains. The protein is localized in the inner membrane and facilitates the rapid transport and exchange of molecules between the cytosol and the mitochondrial matrix space. SLC25A16 gene has a possible role in Graves' disease.This gene encodes a protein that contains three tandemly repeated mitochondrial carrier protein domains. The encoded protein is localized in the inner membrane and facilitates the rapid transport and exchange of molecules between the cytosol and the mitochondrial matrix space.More Product Information