Oakton Electrochemistry Sensors and Electrodes, Cole-Parmer
Supplier: OAKTON INSTRUMENTS MS
WD-35634-50 WD-35650-09 WD-35425-50 WD-35804-00 WD-35808-71
101076-414EA 349.5 USD
101076-414 101076-418 101076-422 101076-432 101076-442
Oakton Electrochemistry Sensors and Electrodes, Cole-Parmer
Electrodes Multi-Parameter Electrodes
Replace sensor modules and electrodes when they can no longer be calibrated, rehydrated, or become contaminated.
Información para pedidos: Select replacement sensor or electrode based on your meter model.