Anti-SLC16A6 Rabbit Polyclonal Antibody
Supplier: Prosci
25-901
10099-760EA 760.88 USD
10099-760
Anti-SLC16A6 Rabbit Polyclonal Antibody
Antibodies
SLC16A6 is a proton-linked monocarboxylate transporter.It catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate.
SLC16A6 antibody can be used for detection of SLC16A6 by ELISA at 1:12500. SLC16A6 antibody can be used for detection of SLC16A6 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinityMore Product Information