Reaction Flask Head, 5-Neck, Scale Up Series™, Ace Glass
Supplier: Ace Glass
6443-02 6443-04 6443-06 6443-08 6443-10 6443-12 6443-14 6443-16 6443-104 6443-106 6443-108 6443-110 6443-112 6443-114 6443-116 6443-118
10098-922EA 534.42 USD
10098-922 10098-924 10098-926 10098-928 10098-930 10098-932 10098-934 10098-936 10098-938 10098-940 10098-942 10098-944 10098-946 10098-948 10098-950 10098-952
Reaction Flask Head, 5-Neck, Scale Up Series™, Ace Glass
Reaction Vessels Reaction Vessels, Flask Shape
Reaction Flasks with 5 neck flat head and flat ground Duran® flange
Flasks each have (1) 90 degree center neck, (1) 90 degree side neck, (2) 10 degree side necks and (1) 45 degree side neck. For use with stainless steel quick release clamps.
Order Now
SPECIFICATIONS
Environmentally Preferable