Anti-GRIA1 Rabbit Polyclonal Antibody
Supplier: Prosci
XPS-1013
10075-662EA 816.2 USD
10075-662
Anti-GRIA1 Rabbit Polyclonal Antibody
Antibodies
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR) while those activated by alpha-amino-3-hydroxy-5-methyl-4-isoxalone propionic acid (AMPA) are known as AMPA receptors (AMPAR). The AMPAR are comprised of 4 distinct subunits (GluR1-4) and they play key roles in virtually all excitatory neurotransmission in the brain. The GluR1 subunit is widely expressed throughout the nervous system. GluR1 is also potentiated by phosphorylation at Ser845. In addition, phosphorylation ofMore Product Information