Human Recombinant Gro Gamma (from E. coli)
Supplier: Prosci
40-393
10072-992EA 305 USD
10072-992
Human Recombinant Gro Gamma (from E. coli)
Proteins and Peptides
All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils. Recombinant human GRO-gamma is a 7.9 kDa protein consisting of 73 amino acids including the 'ELR' motif common to the CXC chemokine family that bind to CXCR1 or CXCR2.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.