Blot Boxes, IBI Scientific
Supplier: IBI Scientific
BLWVS0028 BLWVS0029 ACCBW0013 ACCBW0015 ACCBW0016 ACCBW0017 ACCBW0018 ACCBW0004 ACCBW0010 ACCBW0011 ACCBW0012
10066-576EA 49.3 USD
10066-576 10066-578 10066-580 10066-582 10066-584 10066-586 10066-588 10066-600 10066-602 10066-604 10066-606
Blot Boxes, IBI Scientific
Blotting Apparatus
IBI Blot Boxes are specially designed western blot boxes to fit blots of any size including whole blots, half blots, and strips
The blot boxes are constructed of durable polystyrene plastic with a flat, smooth inner surface and hinged lids to reduce evaporation. The 90⁰ edges create optimal membrane movement for more efficient incubation. With multiple sizes available, you can use a properly sized blot box for your needs to reduce the volume of antibodies required. Available colors include yellow, orange, and clear.
Order Now
SPECIFICATIONS
Environmentally Preferable